Product Details
- SNP ID
-
rs201457055
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:58471047 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACACTCACAGAGCATGCGTTGCT[A/G]GGGAGGCAGCCCAGGACGCCTGAGC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF324
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61742224] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF324
- Gene Name
- zinc finger protein 324
- Gene
- ZNF446
- Gene Name
- zinc finger protein 446
There are no transcripts associated with this gene.
View Full Product Details