Product Details

SNP ID
rs201627212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16576244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGGCTTTGAGGCACCAGCACA[C/T]GCAGCTGCCTGGCCCCTTGCCGGGT
Phenotype
MIM: 605043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED26 PubMed Links

Gene Details

Gene
MED26
Gene Name
mediator complex subunit 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004831.3 1847 Missense Mutation NP_004822.2
Gene
SLC35E1
Gene Name
solute carrier family 35 member E1
There are no transcripts associated with this gene.

View Full Product Details