Product Details

SNP ID
rs200560875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11722020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACTGAAATGACTGAAGGCTTTT[A/C]TACATGTTTTACACTCATAAGGTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF823 PubMed Links

Gene Details

Gene
ZNF823
Gene Name
zinc finger protein 823
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080493.3 1577 Missense Mutation AGA,ATA R505I NP_001073962.1
NM_001297610.1 1577 Missense Mutation AGA,ATA R323I NP_001284539.1
NM_017507.1 1577 Missense Mutation AGA,ATA R461I NP_059977.1
XM_017026941.1 1577 Missense Mutation AGA,ATA R323I XP_016882430.1
XM_017026942.1 1577 Missense Mutation AGA,ATA R323I XP_016882431.1

View Full Product Details