Product Details

SNP ID
rs200595917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11426258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCAAGGTCTCCTCTAGGTACTG[A/C]AGCTGGTTCTGGAGGGCGGGCAGGG
Phenotype
MIM: 615956 MIM: 177060 MIM: 616743
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC151 PubMed Links

Gene Details

Gene
CCDC151
Gene Name
coiled-coil domain containing 151
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302453.1 990 Silent Mutation CTG,CTT L229L NP_001289382.1
NM_001302454.1 990 Silent Mutation CTG,CTT L223L NP_001289383.1
NM_145045.4 990 Silent Mutation CTG,CTT L283L NP_659482.3
XM_017026241.1 990 Silent Mutation CTG,CTT L283L XP_016881730.1
Gene
PRKCSH
Gene Name
protein kinase C substrate 80K-H
There are no transcripts associated with this gene.

Gene
RGL3
Gene Name
ral guanine nucleotide dissociation stimulator like 3
There are no transcripts associated with this gene.

View Full Product Details