Product Details

SNP ID
rs200767135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32692359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCTGCGCCGTGCGGGAGGCCTT[C/T]GAGGAGGCGGGCGTGCTGCTGCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT19 PubMed Links

Gene Details

Gene
NUDT19
Gene Name
nudix hydrolase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105570.1 753 Silent Mutation TTC,TTT F133F NP_001099040.1
XM_017026806.1 753 Silent Mutation TTC,TTT F133F XP_016882295.1

View Full Product Details