Product Details
- SNP ID
-
rs202013490
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:53068462 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAGTCCTTTGATGATTTGCAAGG[C/T]GTGAGTTCTGAGTGAAGACCTTGCC
- Phenotype
-
MIM: 600398
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF160
PubMed Links
Gene Details
- Gene
- ZNF160
- Gene Name
- zinc finger protein 160
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001102603.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001096073.1 |
NM_001322125.1 |
2484 |
Intron |
|
|
NP_001309054.1 |
NM_001322126.1 |
2484 |
Intron |
|
|
NP_001309055.1 |
NM_001322128.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001309057.1 |
NM_001322129.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001309058.1 |
NM_001322130.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001309059.1 |
NM_001322131.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001309060.1 |
NM_001322132.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001309061.1 |
NM_001322133.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001309062.1 |
NM_001322134.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001309063.1 |
NM_001322135.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001309064.1 |
NM_001322136.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_001309065.1 |
NM_001322137.1 |
2484 |
Missense Mutation |
CAC,CGC |
H655R |
NP_001309066.1 |
NM_001322138.1 |
2484 |
Missense Mutation |
CAC,CGC |
H655R |
NP_001309067.1 |
NM_001322139.1 |
2484 |
Missense Mutation |
CAC,CGC |
H655R |
NP_001309068.1 |
NM_033288.3 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_150630.2 |
NM_198893.2 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
NP_942596.1 |
XM_017027445.1 |
2484 |
Missense Mutation |
CAC,CGC |
H710R |
XP_016882934.1 |
XM_017027446.1 |
2484 |
Missense Mutation |
CAC,CGC |
H691R |
XP_016882935.1 |
XM_017027447.1 |
2484 |
Intron |
|
|
XP_016882936.1 |
XM_017027448.1 |
2484 |
Intron |
|
|
XP_016882937.1 |
View Full Product Details