Product Details

SNP ID
rs202121058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15380398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAGCCTCCTCCTGCTCCTTCTC[C/T]TCCTCGGGGCTGTCGGTGAAAGGGT
Phenotype
MIM: 604692 MIM: 609475
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAP8 PubMed Links

Gene Details

Gene
AKAP8
Gene Name
A-kinase anchoring protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005858.3 1668 Intron NP_005849.1
XM_011527624.2 1668 Intron XP_011525926.1
XM_017026141.1 1668 Intron XP_016881630.1
Gene
AKAP8L
Gene Name
A-kinase anchoring protein 8 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291478.1 1668 Silent Mutation NP_001278407.1
NM_014371.3 1668 Silent Mutation NP_055186.3
XM_005259854.3 1668 Missense Mutation XP_005259911.2
XM_011527916.2 1668 Intron XP_011526218.1

View Full Product Details