Product Details

SNP ID
rs202189304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46608902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGTCACGTAATGACGAACCTGGG[A/G]GAGAAGCTGACCGATGAGGAGGTGG
Phenotype
MIM: 114183 MIM: 600022
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALM3 PubMed Links

Gene Details

Gene
CALM3
Gene Name
calmodulin 3
There are no transcripts associated with this gene.

Gene
PPP5D1
Gene Name
PPP5 tetratricopeptide repeat domain containing 1
There are no transcripts associated with this gene.

Gene
PTGIR
Gene Name
prostaglandin I2 (prostacyclin) receptor (IP)
There are no transcripts associated with this gene.

View Full Product Details