Product Details

SNP ID
rs200908461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2933597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTCTCCAGTGTGCGTTCTCAC[A/G]TGCACACGAAGGGACGAGGAACAAC
Phenotype
MIM: 194551
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF77 PubMed Links

Gene Details

Gene
ZNF77
Gene Name
zinc finger protein 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021217.2 1686 Silent Mutation CAC,CAT H510H NP_067040.1
XM_017027081.1 1686 Silent Mutation CAC,CAT H330H XP_016882570.1
XM_017027082.1 1686 Silent Mutation CAC,CAT H330H XP_016882571.1

View Full Product Details