Product Details

SNP ID
rs201665735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52034196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAACATCAGTCCGCTATTCACAA[C/T]GAAACCTTTCCCACATTCACTGCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF432 PubMed Links

Gene Details

Gene
ZNF432
Gene Name
zinc finger protein 432
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322284.1 2105 Missense Mutation ATT,GTT I495V NP_001309213.1
NM_001322285.1 2105 Missense Mutation ATT,GTT I495V NP_001309214.1
NM_014650.3 2105 Missense Mutation ATT,GTT I495V NP_055465.1
Gene
ZNF614
Gene Name
zinc finger protein 614
There are no transcripts associated with this gene.

View Full Product Details