Product Details

SNP ID
rs201114122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:23653293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATGTTTAGTAAGGTTTGCAGAT[C/T]GGTTAAAAGCTTTGTCACATCTCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF675 PubMed Links

Gene Details

Gene
ZNF675
Gene Name
zinc finger protein 675
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138330.2 2440 Intron NP_612203.2
XM_017026442.1 2440 Missense Mutation CAA,CGA Q471R XP_016881931.1
XM_017026443.1 2440 Missense Mutation CAA,CGA Q471R XP_016881932.1
XM_017026444.1 2440 Intron XP_016881933.1

View Full Product Details