Product Details

SNP ID
rs201392131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19529461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCAGAGGAGGAAACAGGCCTGG[A/G]GAAGGCAGTCATGGCTAGAGCAGAT
Phenotype
MIM: 612419 MIM: 609435
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CILP2 PubMed Links

Gene Details

Gene
CILP2
Gene Name
cartilage intermediate layer protein 2
There are no transcripts associated with this gene.

Gene
NDUFA13
Gene Name
NADH:ubiquinone oxidoreductase subunit A13
There are no transcripts associated with this gene.

Gene
YJEFN3
Gene Name
YjeF N-terminal domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190328.1 38 Intron NP_001177257.1
NM_198537.3 38 Missense Mutation AGA,GGA R53G NP_940939.2
XM_005259903.4 38 Missense Mutation AGA,GGA R126G XP_005259960.1
XM_011527996.2 38 Missense Mutation AGA,GGA R126G XP_011526298.1
XM_011527997.1 38 UTR 5 XP_011526299.1
XM_011527998.2 38 Intron XP_011526300.1
XM_011527999.1 38 UTR 5 XP_011526301.1
XM_017026770.1 38 UTR 5 XP_016882259.1
XM_017026771.1 38 UTR 5 XP_016882260.1

View Full Product Details