Product Details

SNP ID
rs201014766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19218930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAACCCAGGCACACAGGATATCA[A/C]CGATGCCAGCGAAAGGGGGCTCCAC
Phenotype
MIM: 600826
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NCAN PubMed Links

Gene Details

Gene
NCAN
Gene Name
neurocan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004386.2 188 Missense Mutation AAC,ACC N30T NP_004377.2

View Full Product Details