Product Details

SNP ID
rs202230938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44703362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTCCTCAGCCACATTCCTGAA[C/T]GTGGGGGCCGAGATCTCTATCACCC
Phenotype
MIM: 614591
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEACAM16 PubMed Links

Gene Details

Gene
CEACAM16
Gene Name
carcinoembryonic antigen related cell adhesion molecule 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039213.3 257 Silent Mutation AAC,AAT N17N NP_001034302.2
XM_017026795.1 257 Silent Mutation AAC,AAT N17N XP_016882284.1

View Full Product Details