Product Details

SNP ID
rs200706703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6306748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTCATCACCCCGGATTTCCACG[C/T]AGGGCAGCCCCACGGGCCAACTGTC
Phenotype
MIM: 613491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACER1 PubMed Links

Gene Details

Gene
ACER1
Gene Name
alkaline ceramidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133492.2 839 Missense Mutation NP_597999.1
XM_011527673.2 839 Missense Mutation XP_011525975.1

View Full Product Details