Product Details

SNP ID
rs201741970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55531150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTCTGCCTACGCACCTTGGGC[C/T]GGATGAAGGCCATGAGGTCCCCGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SBK2 PubMed Links

Gene Details

Gene
SBK2
Gene Name
SH3 domain binding kinase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101401.2 464 Missense Mutation CAG,CGG Q150R NP_001094871.2
XM_006723327.3 464 Missense Mutation CAG,CGG Q150R XP_006723390.1
XM_011527227.2 464 Missense Mutation CAG,CGG Q150R XP_011525529.1
Gene
SBK3
Gene Name
SH3 domain binding kinase family member 3
There are no transcripts associated with this gene.

View Full Product Details