Product Details

SNP ID
rs201813410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51146000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTACCCCTCACAGCCCTCAAA[C/T]CCTCTGGTACTGGAGCTGCAAGTGC
Phenotype
MIM: 604410 MIM: 605640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928517 PubMed Links

Gene Details

Gene
LOC101928517
Gene Name
uncharacterized LOC101928517
There are no transcripts associated with this gene.

Gene
SIGLEC7
Gene Name
sialic acid binding Ig like lectin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277201.1 975 Intron NP_001264130.1
NM_014385.3 975 Silent Mutation AAC,AAT N302N NP_055200.1
NM_016543.3 975 Silent Mutation AAC,AAT N209N NP_057627.2
XM_011526721.2 975 Silent Mutation AAC,AAT N193N XP_011525023.1
Gene
SIGLEC9
Gene Name
sialic acid binding Ig like lectin 9
There are no transcripts associated with this gene.

View Full Product Details