Product Details

SNP ID
rs201082592
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:43997602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTACTGCCCACAATGGGGATCTG[C/T]GCCTTGTCCGGGTGCAGTGGCTGGA
Phenotype
MIM: 604013 MIM: 601019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B4GALT2 PubMed Links

Gene Details

Gene
B4GALT2
Gene Name
beta-1,4-galactosyltransferase 2
There are no transcripts associated with this gene.

Gene
CCDC24
Gene Name
coiled-coil domain containing 24
There are no transcripts associated with this gene.

Gene
SLC6A9
Gene Name
solute carrier family 6 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024845.2 2271 Silent Mutation GCA,GCG A615A NP_001020016.1
NM_001261380.1 2271 Silent Mutation GCA,GCG A619A NP_001248309.1
NM_006934.3 2271 Silent Mutation GCA,GCG A634A NP_008865.2
NM_201649.3 2271 Silent Mutation GCA,GCG A688A NP_964012.2
XM_011542017.2 2271 Intron XP_011540319.1
XM_017002151.1 2271 Silent Mutation GCA,GCG A615A XP_016857640.1
XM_017002152.1 2271 Silent Mutation GCA,GCG A588A XP_016857641.1
XM_017002153.1 2271 Silent Mutation GCA,GCG A577A XP_016857642.1

View Full Product Details