Product Details
- SNP ID
-
rs201322021
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
18
- Location
-
Chr.1:223223456 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCGTGTCCCCTGAGCCGGGGTATG[C/T]TGGGGGGCTCTGGTCGTCCACGGGT
- Phenotype
-
MIM: 615827
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SUSD4
PubMed Links
Gene Details
- Gene
- SUSD4
- Gene Name
- sushi domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037175.2 |
1456 |
Intron |
|
|
NP_001032252.1 |
NM_017982.3 |
1456 |
Missense Mutation |
ACA,GCA |
T413A |
NP_060452.3 |
XM_005273169.1 |
1456 |
Missense Mutation |
ACA,GCA |
T415A |
XP_005273226.1 |
XM_005273172.1 |
1456 |
Missense Mutation |
ACA,GCA |
T253A |
XP_005273229.1 |
XM_006711408.1 |
1456 |
Missense Mutation |
ACA,GCA |
T415A |
XP_006711471.1 |
XM_011509685.1 |
1456 |
Missense Mutation |
ACA,GCA |
T415A |
XP_011507987.1 |
XM_011509687.1 |
1456 |
Missense Mutation |
ACA,GCA |
T255A |
XP_011507989.1 |
XM_017001583.1 |
1456 |
Missense Mutation |
ACA,GCA |
T487A |
XP_016857072.1 |
XM_017001584.1 |
1456 |
Missense Mutation |
ACA,GCA |
T485A |
XP_016857073.1 |
XM_017001585.1 |
1456 |
Missense Mutation |
ACA,GCA |
T472A |
XP_016857074.1 |
XM_017001586.1 |
1456 |
Missense Mutation |
ACA,GCA |
T472A |
XP_016857075.1 |
XM_017001587.1 |
1456 |
Missense Mutation |
ACA,GCA |
T413A |
XP_016857076.1 |
XM_017001588.1 |
1456 |
Missense Mutation |
ACA,GCA |
T413A |
XP_016857077.1 |
XM_017001589.1 |
1456 |
Intron |
|
|
XP_016857078.1 |
View Full Product Details