Product Details

SNP ID
rs201493645
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:86468993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAATGGGGCCATTTAAGAGTTCT[C/G]TGTTCATCTTGATTCTTCACCTTCT
Phenotype
MIM: 603906
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLCA1 PubMed Links

Gene Details

Gene
CLCA1
Gene Name
chloride channel accessory 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285.3 151 Missense Mutation CTG,GTG L8V NP_001276.2

View Full Product Details