Product Details

SNP ID
rs201626667
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:153949262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGAACAAGACTGAGAGGGCAG[A/C]CCTGGCTGCTGTAGAGACTTTGGGG
Phenotype
MIM: 608972 MIM: 604740
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRTC2 PubMed Links

Gene Details

Gene
CRTC2
Gene Name
CREB regulated transcription coactivator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181715.2 1686 Silent Mutation GGG,GGT G509G NP_859066.1
XM_005244946.1 1686 Silent Mutation GGG,GGT G514G XP_005245003.1
XM_005244949.2 1686 Silent Mutation GGG,GGT G401G XP_005245006.1
XM_017000575.1 1686 Silent Mutation GGG,GGT G436G XP_016856064.1
XM_017000576.1 1686 Silent Mutation GGG,GGT G431G XP_016856065.1
XM_017000577.1 1686 Silent Mutation GGG,GGT G413G XP_016856066.1
XM_017000578.1 1686 Silent Mutation GGG,GGT G408G XP_016856067.1
XM_017000579.1 1686 Silent Mutation GGG,GGT G303G XP_016856068.1
Gene
DENND4B
Gene Name
DENN domain containing 4B
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
There are no transcripts associated with this gene.

View Full Product Details