Product Details

SNP ID
rs201353580
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:228106868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGAGCTGAGCCTCACGCTTCCG[C/T]AGCCTGGCCCGGCGCTCCTCAGGAG
Phenotype
MIM: 103180 MIM: 611859
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARF1 PubMed Links

Gene Details

Gene
ARF1
Gene Name
ADP ribosylation factor 1
There are no transcripts associated with this gene.

Gene
C1orf35
Gene Name
chromosome 1 open reading frame 35
There are no transcripts associated with this gene.

Gene
MIR3620
Gene Name
microRNA 3620
There are no transcripts associated with this gene.

Gene
MRPL55
Gene Name
mitochondrial ribosomal protein L55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321284.1 676 Silent Mutation NP_001308213.1
NM_181441.2 676 Silent Mutation NP_852106.1
NM_181454.2 676 Silent Mutation NP_852119.1
NM_181455.2 676 Silent Mutation NP_852120.1
NM_181456.2 676 Silent Mutation NP_852121.1
NM_181462.2 676 Silent Mutation NP_852127.2
NM_181463.2 676 Silent Mutation NP_852128.1
NM_181464.2 676 Silent Mutation NP_852129.1
NM_181465.2 676 Silent Mutation NP_852130.1
XM_005273059.3 676 Silent Mutation XP_005273116.1
XM_005273061.4 676 Silent Mutation XP_005273118.1
XM_005273062.3 676 Silent Mutation XP_005273119.1
XM_005273063.4 676 Silent Mutation XP_005273120.1
XM_011544094.2 676 Silent Mutation XP_011542396.1
XM_011544095.2 676 Silent Mutation XP_011542397.1

View Full Product Details