Product Details
- SNP ID
-
rs201511730
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:92513697 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACGACTCTCTTCTTCTCGGGGGCC[A/G]CTCCTTTTGAACCACTTGGTTGCTC
- Phenotype
-
MIM: 602942
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EVI5
PubMed Links
Gene Details
- Gene
- EVI5
- Gene Name
- ecotropic viral integration site 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308248.1 |
2514 |
Missense Mutation |
CGG,TGG |
R809W |
NP_001295177.1 |
NM_005665.5 |
2514 |
Missense Mutation |
CGG,TGG |
R798W |
NP_005656.4 |
XM_017002269.1 |
2514 |
Missense Mutation |
CGG,TGG |
R918W |
XP_016857758.1 |
XM_017002270.1 |
2514 |
Missense Mutation |
CGG,TGG |
R915W |
XP_016857759.1 |
XM_017002271.1 |
2514 |
Missense Mutation |
CGG,TGG |
R891W |
XP_016857760.1 |
XM_017002272.1 |
2514 |
Missense Mutation |
CGG,TGG |
R885W |
XP_016857761.1 |
XM_017002273.1 |
2514 |
Missense Mutation |
CGG,TGG |
R874W |
XP_016857762.1 |
XM_017002274.1 |
2514 |
Missense Mutation |
CGG,TGG |
R874W |
XP_016857763.1 |
XM_017002275.1 |
2514 |
Missense Mutation |
CGG,TGG |
R874W |
XP_016857764.1 |
XM_017002276.1 |
2514 |
Missense Mutation |
CGG,TGG |
R847W |
XP_016857765.1 |
XM_017002277.1 |
2514 |
Missense Mutation |
CGG,TGG |
R842W |
XP_016857766.1 |
XM_017002278.1 |
2514 |
Missense Mutation |
CGG,TGG |
R836W |
XP_016857767.1 |
XM_017002279.1 |
2514 |
Missense Mutation |
CGG,TGG |
R829W |
XP_016857768.1 |
XM_017002280.1 |
2514 |
Missense Mutation |
CGG,TGG |
R814W |
XP_016857769.1 |
XM_017002281.1 |
2514 |
Missense Mutation |
CGG,TGG |
R806W |
XP_016857770.1 |
XM_017002282.1 |
2514 |
UTR 3 |
|
|
XP_016857771.1 |
XM_017002283.1 |
2514 |
UTR 3 |
|
|
XP_016857772.1 |
XM_017002284.1 |
2514 |
Missense Mutation |
CGG,TGG |
R765W |
XP_016857773.1 |
XM_017002285.1 |
2514 |
UTR 3 |
|
|
XP_016857774.1 |
XM_017002286.1 |
2514 |
Missense Mutation |
CGG,TGG |
R710W |
XP_016857775.1 |
XM_017002287.1 |
2514 |
Missense Mutation |
CGG,TGG |
R710W |
XP_016857776.1 |
XM_017002288.1 |
2514 |
Missense Mutation |
CGG,TGG |
R710W |
XP_016857777.1 |
View Full Product Details