Product Details

SNP ID
rs201314872
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:100874092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCTCAGCTCGATGCCACATTCC[A/G]GAATAGCCACTGTTGGTTTCTTTTT
Phenotype
MIM: 602411
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXTL2 PubMed Links

Gene Details

Gene
EXTL2
Gene Name
exostosin like glycosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033025.2 1770 Silent Mutation TCC,TCT S281S NP_001028197.1
NM_001261440.1 1770 Silent Mutation TCC,TCT S280S NP_001248369.1
NM_001261441.1 1770 Silent Mutation TCC,TCT S289S NP_001248370.1
NM_001261442.1 1770 UTR 3 NP_001248371.1
NM_001439.3 1770 Silent Mutation TCC,TCT S281S NP_001430.1
XM_005270621.1 1770 Silent Mutation TCC,TCT S281S XP_005270678.1
XM_011540994.2 1770 Silent Mutation TCC,TCT S289S XP_011539296.1
XM_011540995.2 1770 Silent Mutation TCC,TCT S289S XP_011539297.1
XM_011540996.2 1770 Silent Mutation TCC,TCT S288S XP_011539298.1
XM_017000651.1 1770 Silent Mutation TCC,TCT S280S XP_016856140.1

View Full Product Details