Product Details

SNP ID
rs201076857
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:153977242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGGAGGCCAGGCCTCCCGGCA[C/T]CCGCAAGCATGGAGCGCCAAGTGTC
Phenotype
MIM: 607138 MIM: 604671 MIM: 602672 MIM: 604740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
There are no transcripts associated with this gene.

Gene
JTB
Gene Name
jumping translocation breakpoint
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006694.3 734 Missense Mutation GAT,GGT D4G NP_006685.1
Gene
RAB13
Gene Name
RAB13, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
There are no transcripts associated with this gene.

View Full Product Details