Product Details

SNP ID
rs201700928
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:225920577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTAAGAGGGTCTTCTTAAGGG[C/T]AGCTGGGGAGATCTTTTCTTGGTCG
Phenotype
MIM: 616406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6741 PubMed Links

Gene Details

Gene
MIR6741
Gene Name
microRNA 6741
There are no transcripts associated with this gene.

Gene
PYCR2
Gene Name
pyrroline-5-carboxylate reductase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271681.1 849 Missense Mutation ACC,GCC T207A NP_001258610.1
NM_013328.3 849 Missense Mutation ACC,GCC T281A NP_037460.2

View Full Product Details