Product Details

SNP ID
rs201013021
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:155248114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTTCCTGGATATCTGGAGGAAA[A/C]GAGTGACCAGGCCCCGACCTGGCCA
Phenotype
MIM: 606463 MIM: 606913
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM189B PubMed Links

Gene Details

Gene
FAM189B
Gene Name
family with sequence similarity 189 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267608.1 2375 Missense Mutation NP_001254537.1
NM_006589.2 2375 Missense Mutation NP_006580.2
NM_198264.1 2375 Missense Mutation NP_937995.1
XM_005244845.1 2375 Missense Mutation XP_005244902.1
Gene
GBA
Gene Name
glucosylceramidase beta
There are no transcripts associated with this gene.

Gene
SCAMP3
Gene Name
secretory carrier membrane protein 3
There are no transcripts associated with this gene.

View Full Product Details