Product Details

SNP ID
rs201240018
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:45018776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGTGCAGTCACTGTGCAGCGT[C/T]GTAAGATGTCTGAAAGCACTGTGGC
Phenotype
MIM: 613521
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HECTD3 PubMed Links

Gene Details

Gene
HECTD3
Gene Name
HECT domain E3 ubiquitin protein ligase 3
There are no transcripts associated with this gene.

Gene
UROD
Gene Name
uroporphyrinogen decarboxylase
There are no transcripts associated with this gene.

Gene
ZSWIM5
Gene Name
zinc finger SWIM-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020883.1 3464 Missense Mutation CAA,CGA Q1079R NP_065934.1
XM_011541861.2 3464 Intron XP_011540163.1
XM_017001913.1 3464 Intron XP_016857402.1

View Full Product Details