Product Details

SNP ID
rs201754622
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:152805151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCCTCCAGAGTGCTGGCCAC[C/T]CCCCCCGCCACAGCAGCTGGAGCCC
Phenotype
MIM: 612604 MIM: 612605 MIM: 612606
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCE1B PubMed Links

Gene Details

Gene
LCE1B
Gene Name
late cornified envelope 1B
There are no transcripts associated with this gene.

Gene
LCE1C
Gene Name
late cornified envelope 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276331.1 289 Missense Mutation AGT,GGT S80G NP_001263260.1
NM_178351.3 289 Missense Mutation AGT,GGT S110G NP_848128.1
Gene
LCE1D
Gene Name
late cornified envelope 1D
There are no transcripts associated with this gene.

View Full Product Details