Product Details
- SNP ID
-
rs201384608
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:247418922 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAAGGGCTGCATCCCCCTCCCGA[A/G]GGGTCAGACAGAGAAGGCAGACCAT
- Phenotype
-
MIM: 606416
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NLRP3
PubMed Links
Gene Details
- Gene
- NLRP3
- Gene Name
- NLR family pyrin domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001079821.2 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
NP_001073289.1 |
NM_001127461.2 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
NP_001120933.1 |
NM_001127462.2 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
NP_001120934.1 |
NM_001243133.1 |
874 |
Missense Mutation |
AAG,AGG |
K41R |
NP_001230062.1 |
NM_004895.4 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
NP_004886.3 |
NM_183395.2 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
NP_899632.1 |
XM_011544048.2 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
XP_011542350.1 |
XM_011544053.2 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
XP_011542355.1 |
XM_011544055.2 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
XP_011542357.1 |
XM_017000181.1 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
XP_016855670.1 |
XM_017000182.1 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
XP_016855671.1 |
XM_017000183.1 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
XP_016855672.1 |
XM_017000184.1 |
874 |
Missense Mutation |
AAG,AGG |
K43R |
XP_016855673.1 |
View Full Product Details