Product Details

SNP ID
rs201610097
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:160681323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGGTGGTTTCAAGCACACTGTT[A/C]TGGAGCTCCTTTGGGAAGGGCCTTT
Phenotype
MIM: 109530
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD48 PubMed Links

Gene Details

Gene
CD48
Gene Name
CD48 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256030.1 687 Missense Mutation CAG,CAT Q177H NP_001242959.1
NM_001778.3 687 Missense Mutation CAG,CAT Q177H NP_001769.2
XM_005245625.1 687 Intron XP_005245682.1
XM_011510171.1 687 Missense Mutation CAG,CAT Q145H XP_011508473.1
XM_017002867.1 687 Missense Mutation CAG,CAT Q76H XP_016858356.1

View Full Product Details