Product Details

SNP ID
rs201095356
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:248522010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTGGCCGTCATGGCTTATGAC[C/T]GCTATGCTGCTGTCTGCCGGCCACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2G6 PubMed Links

Gene Details

Gene
OR2G6
Gene Name
olfactory receptor family 2 subfamily G member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013355.1 364 Missense Mutation CGC,TGC R122C NP_001013373.1
Gene
OR2T10
Gene Name
olfactory receptor family 2 subfamily T member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004693.1 364 Intron NP_001004693.1
XM_017000273.1 364 Intron XP_016855762.1

View Full Product Details