Product Details

SNP ID
rs201426932
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:152566187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTGGGTGGGGGTTGGCACTGCT[G/T]CTGGTTCTGCTGGCAGGACATCTTG
Phenotype
MIM: 612617
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LCE3E PubMed Links

Gene Details

Gene
LCE3E
Gene Name
late cornified envelope 3E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178435.3 59 Missense Mutation AAG,CAG K8Q NP_848522.1
XM_011509484.1 59 Missense Mutation AAG,CAG K8Q XP_011507786.1

View Full Product Details