Product Details

SNP ID
rs199850038
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:155282642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCATCCGCACGCGCTACCTGCG[C/T]ACCTGGTTCCTGGTTGACCTCATCT
Phenotype
MIM: 602989 MIM: 609973 MIM: 609712
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLK2 PubMed Links

Gene Details

Gene
CLK2
Gene Name
CDC like kinase 2
There are no transcripts associated with this gene.

Gene
HCN3
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020897.2 674 Silent Mutation CGC,CGT R170R NP_065948.1
XM_011509816.2 674 Silent Mutation CGC,CGT R123R XP_011508118.1
XM_011509817.2 674 Silent Mutation CGC,CGT R170R XP_011508119.1
XM_011509818.2 674 Intron XP_011508120.1
XM_017001918.1 674 Intron XP_016857407.1
Gene
PKLR
Gene Name
pyruvate kinase, liver and RBC
There are no transcripts associated with this gene.

View Full Product Details