Product Details

SNP ID
rs200070415
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:248239631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACATGGGCTCTGGGGAAAGTCGT[C/T]GCAAGGCCTTCACTACCTGCTCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 703 Missense Mutation CGC,TGC R235C NP_059974.1

View Full Product Details