Product Details

SNP ID
rs199614710
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:23795999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTCTGCTTCTGCCAGCGCCAGA[G/T]ATCCTCACCTGCAACACGGCGAGGT
Phenotype
MIM: 606953 MIM: 613898 MIM: 616143
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GALE PubMed Links

Gene Details

Gene
GALE
Gene Name
UDP-galactose-4-epimerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000403.3 1166 Missense Mutation ATC,CTC I333L NP_000394.2
NM_001008216.1 1166 Missense Mutation ATC,CTC I333L NP_001008217.1
NM_001127621.1 1166 Missense Mutation ATC,CTC I333L NP_001121093.1
Gene
HMGCL
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase
There are no transcripts associated with this gene.

Gene
LYPLA2
Gene Name
lysophospholipase II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007260.2 1166 Intron NP_009191.1
XM_005245728.4 1166 Intron XP_005245785.1
XM_017000165.1 1166 Intron XP_016855654.1
Gene
PITHD1
Gene Name
PITH domain containing 1
There are no transcripts associated with this gene.

View Full Product Details