Product Details

SNP ID
rs199842726
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:226363154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGATATGTGAAGCGTGCTTCAG[C/T]TCATACCTATTCAAAAGAGGACAGT
Phenotype
MIM: 173870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP1 PubMed Links

Gene Details

Gene
PARP1
Gene Name
poly(ADP-ribose) polymerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001618.3 2964 Silent Mutation GAA,GAG E931E NP_001609.2

View Full Product Details