Product Details

SNP ID
rs200082967
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:10463138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGGATGTGGCTCGCCAGCGCAA[C/T]GTCCGAGGAGGTCTCTCTGCTGATA
Phenotype
MIM: 601882
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DFFA PubMed Links

Gene Details

Gene
DFFA
Gene Name
DNA fragmentation factor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004401.2 801 Missense Mutation ATT,GTT I235V NP_004392.1
NM_213566.1 801 Missense Mutation ATT,GTT I235V NP_998731.1

View Full Product Details