Product Details

SNP ID
rs199844062
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:231241435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCGTTGGCCCAACCAGTCCCAG[C/T]GCTGTCGTGCTCCTCTACTCGGTAG
Phenotype
MIM: 602744
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf131 PubMed Links

Gene Details

Gene
C1orf131
Gene Name
chromosome 1 open reading frame 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300830.1 263 Intron NP_001287759.1
NM_152379.3 263 Intron NP_689592.2
Gene
GNPAT
Gene Name
glyceronephosphate O-acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316350.1 263 Silent Mutation AGC,AGT S19S NP_001303279.1
NM_014236.3 263 Silent Mutation AGC,AGT S19S NP_055051.1
XM_005273313.4 263 Silent Mutation AGC,AGT S19S XP_005273370.1
XM_011544303.2 263 UTR 5 XP_011542605.1
XM_011544304.2 263 UTR 5 XP_011542606.1

View Full Product Details