Product Details

SNP ID
rs200101942
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:6192959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGAAAGGCACCTCGGATGTCAC[A/G]GTGATCTTGCTCTTGCTCCTTTCGA
Phenotype
MIM: 180474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL22 PubMed Links

Gene Details

Gene
RPL22
Gene Name
ribosomal protein L22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000983.3 259 Silent Mutation NP_000974.1

View Full Product Details