Product Details
- SNP ID
-
rs199581294
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:152213185 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTCCCCCATCATGGTTACTTCCT[C/T]CTTTGCAATAAGAATACCCATCTTG
- Phenotype
-
MIM: 616293
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HRNR
PubMed Links
Gene Details
- Gene
- HRNR
- Gene Name
- hornerin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001009931.2 |
8523 |
Missense Mutation |
GAA,GGA |
E2815G |
NP_001009931.1 |
View Full Product Details