Product Details

SNP ID
rs199661237
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:204199214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTACCGCACCCAGGAAGGGGATG[A/T]TGCAGACATTGCCCAGGAAGCCGAA
Phenotype
MIM: 603286
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GOLT1A PubMed Links

Gene Details

Gene
GOLT1A
Gene Name
golgi transport 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198447.1 527 Missense Mutation AAC,ATC N114I NP_940849.1
XM_017000314.1 527 Missense Mutation AAC,ATC N114I XP_016855803.1
Gene
KISS1
Gene Name
KiSS-1 metastasis-suppressor
There are no transcripts associated with this gene.

View Full Product Details