Product Details
- SNP ID
-
rs199517077
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:21598052 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACTTGCCGGCGTCTGGGTGGGGT[A/G]ATCGAGAGGGGCCTGGGGAGGGGGG
- Phenotype
-
MIM: 600278
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RAP1GAP
PubMed Links
Gene Details
- Gene
- RAP1GAP
- Gene Name
- RAP1 GTPase activating protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145657.1 |
2150 |
Intron |
|
|
NP_001139129.1 |
NM_001145658.1 |
2150 |
Missense Mutation |
TCA,TTA |
S695L |
NP_001139130.1 |
NM_002885.2 |
2150 |
Missense Mutation |
TCA,TTA |
S631L |
NP_002876.2 |
XM_005245955.3 |
2150 |
Intron |
|
|
XP_005246012.2 |
XM_006710804.2 |
2150 |
Missense Mutation |
TCA,TTA |
S716L |
XP_006710867.2 |
XM_006710805.2 |
2150 |
Intron |
|
|
XP_006710868.2 |
XM_017001965.1 |
2150 |
Missense Mutation |
TCA,TTA |
S716L |
XP_016857454.1 |
XM_017001966.1 |
2150 |
Intron |
|
|
XP_016857455.1 |
XM_017001967.1 |
2150 |
Missense Mutation |
TCA,TTA |
S678L |
XP_016857456.1 |
XM_017001968.1 |
2150 |
Missense Mutation |
TCA,TTA |
S657L |
XP_016857457.1 |
XM_017001969.1 |
2150 |
Missense Mutation |
TCA,TTA |
S657L |
XP_016857458.1 |
XM_017001970.1 |
2150 |
Intron |
|
|
XP_016857459.1 |
XM_017001971.1 |
2150 |
Intron |
|
|
XP_016857460.1 |
XM_017001972.1 |
2150 |
Intron |
|
|
XP_016857461.1 |
XM_017001973.1 |
2150 |
Missense Mutation |
TCA,TTA |
S631L |
XP_016857462.1 |
XM_017001974.1 |
2150 |
Missense Mutation |
TCA,TTA |
S619L |
XP_016857463.1 |
XM_017001975.1 |
2150 |
Intron |
|
|
XP_016857464.1 |
XM_017001976.1 |
2150 |
Missense Mutation |
TCA,TTA |
S593L |
XP_016857465.1 |
XM_017001977.1 |
2150 |
Intron |
|
|
XP_016857466.1 |
XM_017001978.1 |
2150 |
Missense Mutation |
TCA,TTA |
S716L |
XP_016857467.1 |
XM_017001979.1 |
2150 |
Intron |
|
|
XP_016857468.1 |
XM_017001980.1 |
2150 |
Intron |
|
|
XP_016857469.1 |
XM_017001981.1 |
2150 |
Missense Mutation |
TCA,TTA |
S657L |
XP_016857470.1 |
XM_017001982.1 |
2150 |
Missense Mutation |
TCA,TTA |
S657L |
XP_016857471.1 |
XM_017001983.1 |
2150 |
Missense Mutation |
TCA,TTA |
S657L |
XP_016857472.1 |
XM_017001984.1 |
2150 |
Intron |
|
|
XP_016857473.1 |
XM_017001985.1 |
2150 |
Intron |
|
|
XP_016857474.1 |
XM_017001986.1 |
2150 |
Missense Mutation |
TCA,TTA |
S631L |
XP_016857475.1 |
XM_017001987.1 |
2150 |
Missense Mutation |
TCA,TTA |
S631L |
XP_016857476.1 |
XM_017001988.1 |
2150 |
Missense Mutation |
TCA,TTA |
S631L |
XP_016857477.1 |
XM_017001989.1 |
2150 |
Intron |
|
|
XP_016857478.1 |
XM_017001990.1 |
2150 |
Intron |
|
|
XP_016857479.1 |
XM_017001991.1 |
2150 |
Missense Mutation |
TCA,TTA |
S593L |
XP_016857480.1 |
View Full Product Details