Product Details

SNP ID
rs200913655
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.1:107057503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTAGAGCTCTCCCGCGCCGGCT[C/T]GGAGCAGGAGCTGGAGGCCGGAGTG
Phenotype
MIM: 608274
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PRMT6 PubMed Links

Gene Details

Gene
PRMT6
Gene Name
protein arginine methyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018137.2 859 Missense Mutation TCG,TTG S263L NP_060607.2

View Full Product Details