Product Details

SNP ID
rs200498643
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:34761321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTACTCCACAGTGCTGAGCCGC[A/T]TCTGGCTGTCTGTGGTGTTCATCTT
Phenotype
MIM: 605425 MIM: 604493
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GJB4 PubMed Links

Gene Details

Gene
GJB4
Gene Name
gap junction protein beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153212.2 725 Missense Mutation ATC,TTC I23F NP_694944.1
XM_011540679.2 725 Missense Mutation ATC,TTC I23F XP_011538981.1
Gene
GJB5
Gene Name
gap junction protein beta 5
There are no transcripts associated with this gene.

View Full Product Details