Product Details

SNP ID
rs200592125
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:15737938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTGCTCCCATAGATCAAAACG[C/T]AGCTGCAAGCTCAGACAGTGGCCGC
Phenotype
MIM: 609613 MIM: 610817
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHM2 PubMed Links

Gene Details

Gene
PLEKHM2
Gene Name
pleckstrin homology and RUN domain containing M2
There are no transcripts associated with this gene.

Gene
SLC25A34
Gene Name
solute carrier family 25 member 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207348.2 1786 Nonsense Mutation CAG,TAG Q130* NP_997231.1
XM_011541292.2 1786 Nonsense Mutation CAG,TAG Q130* XP_011539594.1
XM_011541293.1 1786 Nonsense Mutation CAG,TAG Q130* XP_011539595.1
XM_017001083.1 1786 Nonsense Mutation CAG,TAG Q130* XP_016856572.1
Gene
TMEM82
Gene Name
transmembrane protein 82
There are no transcripts associated with this gene.

View Full Product Details