Product Details
- SNP ID
-
rs200500421
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:201361337 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCGAACTTCTCTGCCTCCAAGTTA[C/T]AGATGCTCTGCCACAGCTCCTTGGC
- Phenotype
-
MIM: 191045
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TNNT2
PubMed Links
Gene Details
- Gene
- TNNT2
- Gene Name
- troponin T2, cardiac type
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000364.3 |
939 |
Missense Mutation |
TAT,TGT |
Y248C |
NP_000355.2 |
NM_001001430.2 |
939 |
Missense Mutation |
TAT,TGT |
Y241C |
NP_001001430.1 |
NM_001001431.2 |
939 |
Missense Mutation |
TAT,TGT |
Y238C |
NP_001001431.1 |
NM_001001432.2 |
939 |
Missense Mutation |
TAT,TGT |
Y235C |
NP_001001432.1 |
NM_001276345.1 |
939 |
Missense Mutation |
TAT,TGT |
Y251C |
NP_001263274.1 |
NM_001276346.1 |
939 |
Missense Mutation |
TAT,TGT |
Y208C |
NP_001263275.1 |
NM_001276347.1 |
939 |
Missense Mutation |
TAT,TGT |
Y241C |
NP_001263276.1 |
XM_006711508.3 |
939 |
Missense Mutation |
TAT,TGT |
Y241C |
XP_006711571.1 |
XM_006711509.3 |
939 |
Missense Mutation |
TAT,TGT |
Y240C |
XP_006711572.1 |
XM_011509938.2 |
939 |
Missense Mutation |
TAT,TGT |
Y251C |
XP_011508240.1 |
XM_011509939.1 |
939 |
Missense Mutation |
TAT,TGT |
Y250C |
XP_011508241.1 |
XM_011509940.2 |
939 |
Missense Mutation |
TAT,TGT |
Y250C |
XP_011508242.1 |
XM_011509941.2 |
939 |
Missense Mutation |
TAT,TGT |
Y249C |
XP_011508243.1 |
XM_011509942.2 |
939 |
Missense Mutation |
TAT,TGT |
Y236C |
XP_011508244.1 |
XM_011509943.2 |
939 |
Missense Mutation |
TAT,TGT |
Y236C |
XP_011508245.1 |
XM_011509944.2 |
939 |
Missense Mutation |
TAT,TGT |
Y235C |
XP_011508246.1 |
XM_011509946.1 |
939 |
Missense Mutation |
TAT,TGT |
Y182C |
XP_011508248.1 |
XM_017002216.1 |
939 |
Missense Mutation |
TAT,TGT |
Y240C |
XP_016857705.1 |
XM_017002217.1 |
939 |
Missense Mutation |
TAT,TGT |
Y238C |
XP_016857706.1 |
View Full Product Details