Product Details

SNP ID
rs200833497
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:167810892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAGACAGGGCCAGTGGTATTTT[C/G]AGCCACCAGATTCTCCAAGTTCTAA
Phenotype
MIM: 605205
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADCY10 PubMed Links

Gene Details

Gene
ADCY10
Gene Name
adenylate cyclase 10, soluble
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167749.2 5981 Missense Mutation CAA,GAA Q1349E NP_001161221.1
NM_001297772.1 5981 Missense Mutation CAA,GAA Q1410E NP_001284701.1
NM_018417.5 5981 Missense Mutation CAA,GAA Q1502E NP_060887.2
XM_005245330.4 5981 Intron XP_005245387.1
XM_006711449.3 5981 Intron XP_006711512.1
XM_011509760.2 5981 Missense Mutation CAA,GAA Q1502E XP_011508062.1
XM_011509762.2 5981 Missense Mutation CAA,GAA Q1454E XP_011508064.1
XM_011509763.2 5981 Missense Mutation CAA,GAA Q1448E XP_011508065.1
XM_011509766.2 5981 Intron XP_011508068.1
XM_017001778.1 5981 Intron XP_016857267.1

View Full Product Details