Product Details

SNP ID
rs200249782
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:22659556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCCCAGCTCAGCTGCACCGGG[C/T]CCCCAGCCATCCCTGGCATCCCGGG
Phenotype
MIM: 120570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QB PubMed Links

Gene Details

Gene
C1QB
Gene Name
complement C1q B chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000491.3 232 Missense Mutation CCC,TCC P34S NP_000482.3
XM_011542059.2 232 Missense Mutation CCC,TCC P34S XP_011540361.1

View Full Product Details