Product Details

SNP ID
rs200595214
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:248239320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTGGCTATATGTCACCCTCTTC[A/G]GTACACCATCCTCATGAATCCGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 392 Missense Mutation CAG,CGG Q131R NP_059974.1

View Full Product Details